Learn More
Immunology Consultants anti-Bovine Rotavirus Antibody (Mouse - Clone #101) - Monoclonal
SDP
Catalog No. 5000435340
Encompass_Preferred
Clone 101 targets a distinct epitope on bovine rotavirus, providing complementary detection to Clone 100. This monoclonal antibody supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and low background. MRTV‑10A‑101 is well suited for diagnostic assay development, surveillance studies, and research examining viral infection dynamics in calves. Its reproducible performance ensures dependable detection across fecal extracts, culture fluids, and purified antigen preparations, making it a robust reagent for veterinary virology applications.
Catalog No. 50-004-35340 Supplier Immunology Consultants Supplier No. MRTV10A101
Clone 101 targets a distinct epitope on bovine rotavirus, providing complementary detection to Clone 100. This monoclonal antibody supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and low background. MRTV‑10A‑101 is well suited for diagnostic assay development, surveillance studies, and research examining viral infection dynamics in calves. Its reproducible performance ensures dependable detection across fecal extracts, culture fluids, and purified antigen preparations, making it a robust reagent for veterinary virology applications.