Learn More
Immunology Consultants anti-Bovine SAA Antibody (Mouse - Clone #1F12) - Monoclonal
SDP
Catalog No. 5000435423
Encompass_Preferred
This monoclonal antibody provides sensitive, specific detection of bovine Serum Amyloid A (SAA), a key acute‑phase protein used to monitor inflammation and disease in cattle. Clone 1F12 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture assays, enabling precise quantification of SAA in serum, plasma, and purified antigen samples. MSAA‑10A‑1F12 is ideal for herd‑health monitoring, diagnostic assay development, and research on bovine inflammatory responses. Its consistent lot‑to‑lot performance ensures reliable detection across repeated assays.
Catalog No. 50-004-35423 Supplier Immunology Consultants Supplier No. MSAA10A1F12
This monoclonal antibody provides sensitive, specific detection of bovine Serum Amyloid A (SAA), a key acute‑phase protein used to monitor inflammation and disease in cattle. Clone 1F12 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture assays, enabling precise quantification of SAA in serum, plasma, and purified antigen samples. MSAA‑10A‑1F12 is ideal for herd‑health monitoring, diagnostic assay development, and research on bovine inflammatory responses. Its consistent lot‑to‑lot performance ensures reliable detection across repeated assays.