Learn More
Immunology Consultants anti-BSA Antibody (Rabbit) - Affinity Purified
SDP
Catalog No. 5000435503
Encompass_Preferred
RAL‑10A is an affinity‑purified rabbit polyclonal antibody targeting bovine serum albumin (BSA). This preparation offers enhanced specificity and reduced background compared to whole serum, making it ideal for sensitive immunoassays and applications requiring high signal‑to‑noise performance. The purification process enriches for high‑affinity immunoglobulins, ensuring consistent binding characteristics and improved reproducibility across experiments. Produced by Immunology Consultants, this antibody is well suited for ELISA, Western blotting, immunoprecipitation, and other protein detection workflows. Supplied in a 1 mg format, RAL‑10A provides a reliable, high‑quality reagent for researchers working with albumin‑based assays or studying albumin‑related biological processes.
Catalog No. 50-004-35503 Supplier Immunology Consultants Supplier No. RAL10A
RAL‑10A is an affinity‑purified rabbit polyclonal antibody targeting bovine serum albumin (BSA). This preparation offers enhanced specificity and reduced background compared to whole serum, making it ideal for sensitive immunoassays and applications requiring high signal‑to‑noise performance. The purification process enriches for high‑affinity immunoglobulins, ensuring consistent binding characteristics and improved reproducibility across experiments. Produced by Immunology Consultants, this antibody is well suited for ELISA, Western blotting, immunoprecipitation, and other protein detection workflows. Supplied in a 1 mg format, RAL‑10A provides a reliable, high‑quality reagent for researchers working with albumin‑based assays or studying albumin‑related biological processes.