Learn More
Immunology Consultants anti-BSA Antibody (Rabbit) - IgG Fraction
SDP
Catalog No. 5000435402
Encompass_Preferred
RAL‑10GLY is a rabbit anti‑BSA antibody supplied as an isolated IgG fraction, offering improved purity and reduced non‑IgG serum components. This format provides consistent performance in assays requiring defined immunoglobulin content, such as quantitative ELISA, Western blotting, and immunochemical detection of albumin. The IgG fractionation process enhances assay reproducibility and minimizes background interference, making it particularly valuable for high‑precision workflows. Manufactured by Immunology Consultants, this reagent delivers strong, specific binding to bovine serum albumin and is supplied in a 50 mg quantity, supporting extensive experimental use. RAL‑10GLY is an excellent choice for laboratories requiring a purified, high‑performance anti‑BSA antibody.
Catalog No. 50-004-35402 Supplier Immunology Consultants Supplier No. RAL10GLY
RAL‑10GLY is a rabbit anti‑BSA antibody supplied as an isolated IgG fraction, offering improved purity and reduced non‑IgG serum components. This format provides consistent performance in assays requiring defined immunoglobulin content, such as quantitative ELISA, Western blotting, and immunochemical detection of albumin. The IgG fractionation process enhances assay reproducibility and minimizes background interference, making it particularly valuable for high‑precision workflows. Manufactured by Immunology Consultants, this reagent delivers strong, specific binding to bovine serum albumin and is supplied in a 50 mg quantity, supporting extensive experimental use. RAL‑10GLY is an excellent choice for laboratories requiring a purified, high‑performance anti‑BSA antibody.