Learn More
Immunology Consultants anti-BSA Antibody (Sheep) - HRP Conjugated
SDP
Catalog No. 5000435425
Encompass_Preferred
This HRP‑conjugated sheep anti‑BSA antibody enables sensitive enzymatic detection of bovine serum albumin in ELISA and Western blotting. The HRP label provides strong signal amplification, supporting high‑contrast, low‑background assay results. Manufactured by Immunology Consultants, this 1 mg reagent is ideal for quality‑control testing, contamination monitoring, and albumin‑based assay development.
Catalog No. 50-004-35425 Supplier Immunology Consultants Supplier No. SAL10P
This HRP‑conjugated sheep anti‑BSA antibody enables sensitive enzymatic detection of bovine serum albumin in ELISA and Western blotting. The HRP label provides strong signal amplification, supporting high‑contrast, low‑background assay results. Manufactured by Immunology Consultants, this 1 mg reagent is ideal for quality‑control testing, contamination monitoring, and albumin‑based assay development.