Learn More
Immunology Consultants anti-BSA (Rabbit) - Whole Serum
SDP
Catalog No. 5000435487
Encompass_Preferred
RAL‑10 is a rabbit-derived anti-BSA primary antibody supplied as whole antiserum. This reagent is designed for applications requiring broad reactivity to bovine serum albumin, making it suitable for immunodetection, assay development, and protein quantification workflows. As a whole serum preparation, it contains the full complement of polyclonal antibodies generated during immunization, offering strong signal and robust binding across multiple assay formats. Produced by Immunology Consultants, this antibody provides dependable performance in ELISA, Western blotting, and other immunochemical techniques where albumin detection is required. Supplied in a 1 mL format, it is an economical and versatile choice for laboratories needing consistent anti-BSA activity.
Catalog No. 50-004-35487 Supplier Immunology Consultants Supplier No. RAL10
Add to Cart
Add to Cart
RAL‑10 is a rabbit-derived anti-BSA primary antibody supplied as whole antiserum. This reagent is designed for applications requiring broad reactivity to bovine serum albumin, making it suitable for immunodetection, assay development, and protein quantification workflows. As a whole serum preparation, it contains the full complement of polyclonal antibodies generated during immunization, offering strong signal and robust binding across multiple assay formats. Produced by Immunology Consultants, this antibody provides dependable performance in ELISA, Western blotting, and other immunochemical techniques where albumin detection is required. Supplied in a 1 mL format, it is an economical and versatile choice for laboratories needing consistent anti-BSA activity.