Learn More
Eurofins Discoverx Anti-BTLA Antibody [6F4], 100 µg. (92-1332)
Supplier:  Eurofins Discoverx 921332
Encompass
Human BTLA (or B and T lymphocyte associated), is an Ig domain superfamily protein with cytoplasmic immunoreceptor tyrosine-based inhibitory motif that is induced during activation of T cells. The herpes virus entry mediator (HVEM), a member of the TNF receptor (TNFR) superfamily, can act as a molecular switch that modulates T cell activation by propagating positive signals from the TNF-related ligand LIGHT (TNFR superfamily 14), or inhibitory signals through the Ig superfamily member BTLA. The binding site on HVEM for BTLA is conserved in the orphan TNF receptor UL144, present in human CMV. Provided as 100 μg of antibody in 100 μL buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9842