Learn More
Immunology Consultants anti-C-Myc Antibody (Chicken) - Affinity Purified
SDP
Catalog No. 5000435361
Encompass_Preferred
This 0.1 mg format delivers the same high‑performance, affinity‑purified chicken antibody used in CMYC‑45A, optimized for laboratories needing smaller quantities for pilot studies, low‑volume assays, or targeted validation work. The antibody provides strong, specific recognition of the C‑Myc epitope tag with minimal cross‑reactivity in mammalian systems, supporting clean detection in ELISA, Western blotting, and fluorescence‑based applications. Its avian host species and stringent purification contribute to low background and consistent assay behavior, making this economical vial size a practical option for method development or limited‑scope experiments involving C‑Myc–tagged proteins.
Catalog No. 50-004-35361 Supplier Immunology Consultants Supplier No. CMYC45AZ
This 0.1 mg format delivers the same high‑performance, affinity‑purified chicken antibody used in CMYC‑45A, optimized for laboratories needing smaller quantities for pilot studies, low‑volume assays, or targeted validation work. The antibody provides strong, specific recognition of the C‑Myc epitope tag with minimal cross‑reactivity in mammalian systems, supporting clean detection in ELISA, Western blotting, and fluorescence‑based applications. Its avian host species and stringent purification contribute to low background and consistent assay behavior, making this economical vial size a practical option for method development or limited‑scope experiments involving C‑Myc–tagged proteins.