Learn More
Immunology Consultants anti-C-Myc Antibody (Chicken) - Biotin Conjugated
SDP
Catalog No. 5000435362
Encompass_Preferred
This biotin‑conjugated chicken antibody enables flexible, high‑sensitivity detection of C‑Myc–tagged proteins through streptavidin‑based amplification systems. Ideal for ELISA, Western blotting, and solid‑phase assays, the biotin label supports enhanced signal generation while maintaining the strong specificity and low background associated with avian antibodies. The conjugate is produced from affinity‑purified material to ensure consistent performance and minimal nonspecific binding. CMYC‑45B is well suited for workflows requiring modular detection strategies, high‑throughput assay development, or applications where biotin–streptavidin chemistry provides superior sensitivity and dynamic range.
Catalog No. 50-004-35362 Supplier Immunology Consultants Supplier No. CMYC45B
This biotin‑conjugated chicken antibody enables flexible, high‑sensitivity detection of C‑Myc–tagged proteins through streptavidin‑based amplification systems. Ideal for ELISA, Western blotting, and solid‑phase assays, the biotin label supports enhanced signal generation while maintaining the strong specificity and low background associated with avian antibodies. The conjugate is produced from affinity‑purified material to ensure consistent performance and minimal nonspecific binding. CMYC‑45B is well suited for workflows requiring modular detection strategies, high‑throughput assay development, or applications where biotin–streptavidin chemistry provides superior sensitivity and dynamic range.