Learn More
Immunology Consultants anti-C-Myc Antibody (Chicken) - HRP Conjugated
SDP
Catalog No. 5000435449
Encompass_Preferred
This HRP‑conjugated chicken antibody provides direct, enzyme‑based detection of C‑Myc–tagged proteins, eliminating the need for secondary antibodies and reducing assay complexity. The conjugate delivers strong, reliable signal in chemiluminescent and colorimetric ELISA and Western blot formats, supported by the low background inherent to avian antibodies. Produced from affinity‑purified material, it offers consistent specificity and robust performance even in complex mammalian samples. The 0.1 mg vial size is ideal for small‑scale studies, rapid screening, or assay optimization requiring precise, HRP‑based detection of C‑Myc epitopes.
Catalog No. 50-004-35449 Supplier Immunology Consultants Supplier No. CMYC45PZ
This HRP‑conjugated chicken antibody provides direct, enzyme‑based detection of C‑Myc–tagged proteins, eliminating the need for secondary antibodies and reducing assay complexity. The conjugate delivers strong, reliable signal in chemiluminescent and colorimetric ELISA and Western blot formats, supported by the low background inherent to avian antibodies. Produced from affinity‑purified material, it offers consistent specificity and robust performance even in complex mammalian samples. The 0.1 mg vial size is ideal for small‑scale studies, rapid screening, or assay optimization requiring precise, HRP‑based detection of C‑Myc epitopes.