Learn More
Immunology Consultants anti-C-Myc Antibody (Goat) - Affinity Purified
SDP
Catalog No. 5000435498
Encompass_Preferred
This 0.1 mg format provides the same high‑specificity detection of the C‑Myc epitope tag as GMYC‑45A, optimized for pilot studies, assay development, and low‑volume workflows. The antibody supports ELISA, Western blotting, and protein characterization with strong affinity and low background. GMYC‑45A‑Z is ideal for method optimization, small‑scale validation, and applications requiring consistent, reproducible detection of C‑Myc–tagged proteins.
Catalog No. 50-004-35498 Supplier Immunology Consultants Supplier No. GMYC45AZ
This 0.1 mg format provides the same high‑specificity detection of the C‑Myc epitope tag as GMYC‑45A, optimized for pilot studies, assay development, and low‑volume workflows. The antibody supports ELISA, Western blotting, and protein characterization with strong affinity and low background. GMYC‑45A‑Z is ideal for method optimization, small‑scale validation, and applications requiring consistent, reproducible detection of C‑Myc–tagged proteins.