Learn More
Immunology Consultants anti-C-MYC Antibody (Mouse - Clone #7C12) - Monoclonal
SDP
Catalog No. 5000435456
Encompass_Preferred
This monoclonal antibody provides highly specific detection of the C‑Myc epitope tag, widely used in recombinant protein expression and purification. Clone 7C12 delivers strong affinity and low background in ELISA, Western blotting, immunoprecipitation, and protein‑tracking workflows. Its clean, reproducible signal enables precise monitoring of Myc‑tagged proteins in cell lysates, culture supernatants, and purified preparations. MMYC‑45A‑7C12 is ideal for molecular biology, protein engineering, and assay development requiring reliable Myc‑tag detection.
Catalog No. 50-004-35456 Supplier Immunology Consultants Supplier No. MMYC45A7C12
This monoclonal antibody provides highly specific detection of the C‑Myc epitope tag, widely used in recombinant protein expression and purification. Clone 7C12 delivers strong affinity and low background in ELISA, Western blotting, immunoprecipitation, and protein‑tracking workflows. Its clean, reproducible signal enables precise monitoring of Myc‑tagged proteins in cell lysates, culture supernatants, and purified preparations. MMYC‑45A‑7C12 is ideal for molecular biology, protein engineering, and assay development requiring reliable Myc‑tag detection.