Learn More
Immunology Consultants anti-C-MYC Antibody (Mouse - Clone #7C12) - Monoclonal
SDP
Catalog No. 5000435478
Encompass_Preferred
This 0.1 mg format provides the same high‑specificity detection of the C‑Myc epitope tag as MMYC‑45A‑7C12, optimized for small‑scale workflows, pilot studies, and assay development. Clone 7C12 offers strong affinity and low background in ELISA, Western blotting, and protein‑characterization assays. MMYC‑45A‑7C12‑Z is ideal for laboratories requiring reliable Myc‑tag detection in low‑volume applications, providing consistent, reproducible performance across diverse sample types.
Catalog No. 50-004-35478 Supplier Immunology Consultants Supplier No. MMYC45A7C12Z
This 0.1 mg format provides the same high‑specificity detection of the C‑Myc epitope tag as MMYC‑45A‑7C12, optimized for small‑scale workflows, pilot studies, and assay development. Clone 7C12 offers strong affinity and low background in ELISA, Western blotting, and protein‑characterization assays. MMYC‑45A‑7C12‑Z is ideal for laboratories requiring reliable Myc‑tag detection in low‑volume applications, providing consistent, reproducible performance across diverse sample types.