Learn More
Immunology Consultants anti-C-MYC Antibody (Mouse - Clone #8F8) - Monoclonal
SDP
Catalog No. 5000435337
Encompass_Preferred
This 0.1 mg format provides the same high‑specificity Myc‑tag detection as MMYC‑45A‑8F8, optimized for low‑volume workflows and pilot‑scale assay development. Clone 8F8 delivers strong affinity and clean signal in ELISA, Western blotting, and immunoprecipitation. MMYC‑45A‑8F8‑Z offers consistent, reproducible performance for laboratories requiring small‑format Myc‑tag reagents.
Catalog No. 50-004-35337 Supplier Immunology Consultants Supplier No. MMYC45A8F8Z
This 0.1 mg format provides the same high‑specificity Myc‑tag detection as MMYC‑45A‑8F8, optimized for low‑volume workflows and pilot‑scale assay development. Clone 8F8 delivers strong affinity and clean signal in ELISA, Western blotting, and immunoprecipitation. MMYC‑45A‑8F8‑Z offers consistent, reproducible performance for laboratories requiring small‑format Myc‑tag reagents.