Learn More
Immunology Consultants anti-C-MYC Antibody (Mouse - Clone #8F8) - Monoclonal
SDP
Catalog No. 5000435420
Encompass_Preferred
Clone 8F8 targets a distinct epitope on the C‑Myc tag, providing complementary detection to Clone 7C12. This monoclonal antibody supports sensitive measurement in ELISA, Western blotting, immunoprecipitation, and protein‑tracking workflows. Its strong affinity and minimal nonspecific binding enable precise detection of Myc‑tagged proteins in lysates, supernatants, and purified preparations. MMYC‑45A‑8F8 is ideal for molecular biology, recombinant protein characterization, and assay development requiring robust Myc‑tag recognition.
Catalog No. 50-004-35420 Supplier Immunology Consultants Supplier No. MMYC45A8F8
Clone 8F8 targets a distinct epitope on the C‑Myc tag, providing complementary detection to Clone 7C12. This monoclonal antibody supports sensitive measurement in ELISA, Western blotting, immunoprecipitation, and protein‑tracking workflows. Its strong affinity and minimal nonspecific binding enable precise detection of Myc‑tagged proteins in lysates, supernatants, and purified preparations. MMYC‑45A‑8F8 is ideal for molecular biology, recombinant protein characterization, and assay development requiring robust Myc‑tag recognition.