Learn More
Immunology Consultants anti-C-Myc Antibody (Rabbit) - Affinity Purified
SDP
Catalog No. 5000435468
Encompass_Preferred
This 0.1 mg affinity‑purified anti‑C‑Myc antibody provides highly specific detection of C‑Myc–tagged proteins in Western blotting, ELISA, immunofluorescence, and immunoprecipitation. Ideal for limited‑sample experiments or assay development, it delivers strong epitope recognition with minimal background. Produced by Immunology Consultants, the antibody’s clean binding profile ensures consistent performance across diverse assay formats. Its small size makes it a practical choice for researchers optimizing detection conditions or evaluating C‑Myc–tagged constructs without requiring a full‑scale reagent quantity.
Catalog No. 50-004-35468 Supplier Immunology Consultants Supplier No. RMYC45AZ
This 0.1 mg affinity‑purified anti‑C‑Myc antibody provides highly specific detection of C‑Myc–tagged proteins in Western blotting, ELISA, immunofluorescence, and immunoprecipitation. Ideal for limited‑sample experiments or assay development, it delivers strong epitope recognition with minimal background. Produced by Immunology Consultants, the antibody’s clean binding profile ensures consistent performance across diverse assay formats. Its small size makes it a practical choice for researchers optimizing detection conditions or evaluating C‑Myc–tagged constructs without requiring a full‑scale reagent quantity.