Learn More
Immunology Consultants anti-C-Myc Antibody (Rabbit) - HRP Conjugated
SDP
Catalog No. 5000435352
Encompass_Preferred
This 0.1 mg HRP‑conjugated anti‑C‑Myc antibody provides powerful enzymatic detection in a compact, flexible format ideal for small‑scale studies or assay optimization. The HRP label supports strong chemiluminescent or colorimetric signal generation, enabling sensitive detection of C‑Myc–tagged proteins in Western blotting and ELISA. Manufactured by Immunology Consultants, the antibody maintains high affinity for the C‑Myc epitope and delivers reliable, low‑background performance. Its smaller size is well suited for pilot experiments or workflows requiring precise, enzyme‑linked detection
Catalog No. 50-004-35352 Supplier Immunology Consultants Supplier No. RMYC45PZ
This 0.1 mg HRP‑conjugated anti‑C‑Myc antibody provides powerful enzymatic detection in a compact, flexible format ideal for small‑scale studies or assay optimization. The HRP label supports strong chemiluminescent or colorimetric signal generation, enabling sensitive detection of C‑Myc–tagged proteins in Western blotting and ELISA. Manufactured by Immunology Consultants, the antibody maintains high affinity for the C‑Myc epitope and delivers reliable, low‑background performance. Its smaller size is well suited for pilot experiments or workflows requiring precise, enzyme‑linked detection