Learn More
Immunology Consultants anti-C-Myc Antibody (Rabbit) - HRP Conjugated
SDP
Supplier:  Immunology Consultants RMYC45P
Encompass_Preferred
This HRP‑conjugated anti‑C‑Myc antibody provides sensitive enzymatic detection of C‑Myc–tagged proteins in Western blotting, ELISA, and chemiluminescent or colorimetric assays. The HRP label enables strong signal amplification, making it ideal for detecting low‑abundance proteins or achieving high‑contrast results. Produced by Immunology Consultants, this 1 mg reagent offers excellent specificity and consistent performance across assay platforms. It is a dependable choice for researchers requiring robust, enzyme‑linked detection of C‑Myc–tagged constructs in molecular biology and protein analysis workflows
Catalog No. 50-004-35385