Learn More
Immunology Consultants FITC Conjugated Chicken anti-C-Myc Antibody, 1.0 mg
SDP
Supplier:  Immunology Consultants CMYC45F
Encompass_Preferred
Chickens were immunized with EQKLISEEDL (C-MYC) conjugated with KLH. Eggs were collected and the Ig fraction prepared. Antibodies were immunoaffinity purified using the peptide immobilized on a solid phase matrix then conjugated with fluorescein isothiocyanate (FITC). Antibody concentration was determined using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. FITC/Protein Ratio = 3.0 –6.0. This antibody will react with EQKLISEEDL as determined by ELISA and IEP techniques. It is suitable for blotting, ELISA and IF applications. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. NC1749149