Learn More
Sino Biological Anti-C-Reactive Protein Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000786396
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat C-Reactive Protein (rR C-Reactive Protein; Catalog#80041-R08H; NP_058792.1; Met1-Ser230). C-Reactive Protein specific IgG was purified by Rat C-Reactive Protein affinity chromatography.
Catalog No. 50-007-86396 Supplier Sino Biological Supplier No. 80041T32100
Produced in rabbits immunized with purified, recombinant Rat C-Reactive Protein (rR C-Reactive Protein; Catalog#80041-R08H; NP_058792.1; Met1-Ser230). C-Reactive Protein specific IgG was purified by Rat C-Reactive Protein affinity chromatography.