Learn More
ABclonal Technology C4BPA Polyclonal Antibody, 100UL, Validated in:WB, IHC
SDP
Supplier:  ABclonal Technology A7648
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to a sequence within amino acids 500 to the C-terminus of human C4BPA (NP_000706.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC•Swiss Prot: P04003•Gene ID:722•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-9119