Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-CA125, 0.2mg
Supplier: Advanced ImmunoChemical, Inc. 2CA1250.2MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal antibodies / Tumor Markers / Monoclonal mouse anti-CA125. MAbs:X75, X306, X325, X52Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with CA125 antigen purified from human ovarian carcinoma (MW > 1 MDa).Specificity:CA125 antigen. MAbs X52, X75, X325: epitope specificity group B (ISOBM classification) similar to M-11.MAb X306: epitope specificity group A (ISOBM classification) similar to OC125.MAb Isotypes:IgG1 for all MAbsApplications:Detection of CA125 antigen. Suggested pairs for sandwich immunoassay (capture-detection):X306 – X52X306 – X325X306 – X75All MAbs are working in Western blotting.paraffin embedded tissue.Purification:MIon exchange chromatographyPresentation:MAb 10C7 in PBS, pH 7.4, 0.1 % sodium azide (NaN3)MAbs TSB1 and TSB4 in 0.15 M NaCl, 10 mM Tris, pH 7.5, 0.05% sodium azide (NaN3)Storage:+ 4 °C