Learn More
Sino Biological Anti-Calreticulin/CALR Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000748990
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Calreticulin/CALR (rh Calreticulin/CALR; Catalog#13539-H08H; P27797; Met1-Ala413). Calreticulin/CALR specific IgG was purified by Human Calreticulin/CALR affinity chromatography.
Catalog No. 50-007-48990 Supplier Sino Biological Supplier No. 13539T54100
Produced in rabbits immunized with purified, recombinant Human Calreticulin/CALR (rh Calreticulin/CALR; Catalog#13539-H08H; P27797; Met1-Ala413). Calreticulin/CALR specific IgG was purified by Human Calreticulin/CALR affinity chromatography.