Learn More
Sino Biological Anti-CAMKIV Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000786579
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CAMKIV (rM CAMKIV; Catalog#50838-M20B; P08414; Met1-Tyr469). CAMKIV specific IgG was purified by Mouse CAMKIV affinity chromatography.
Catalog No. 50-007-86579 Supplier Sino Biological Supplier No. 50838T24100
Produced in rabbits immunized with purified, recombinant Mouse CAMKIV (rM CAMKIV; Catalog#50838-M20B; P08414; Met1-Tyr469). CAMKIV specific IgG was purified by Mouse CAMKIV affinity chromatography.