Learn More
Immunology Consultants Affinity Purified Chicken anti-Campylobacter Jejuni Antibody Clone #202, 1.0 mg
SDP
Catalog No. NC2734239
Encompass_Preferred
Chickens were immunized with highly purified Campylobacter jejuni hippurate hydrolase (HipO) and the eggs were collected and delipidated. Antibody was immunoaffinity purified off a HipO containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. This antibody reacts with Campylobacter jejuni hippurate hydrolase as determined by ELISA and it can detect Campylobacter jejuni cells with Immunofluorescence staining. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. NC2734239 Supplier Immunology Consultants Supplier No. CCBJ55A202
Chickens were immunized with highly purified Campylobacter jejuni hippurate hydrolase (HipO) and the eggs were collected and delipidated. Antibody was immunoaffinity purified off a HipO containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. This antibody reacts with Campylobacter jejuni hippurate hydrolase as determined by ELISA and it can detect Campylobacter jejuni cells with Immunofluorescence staining. Optimal working dilutions should be determined experimentally by the investigator.