Learn More
Seracare Life Sciences Inc ANTI-CAMPYLOBACTER SPECIES AB
Supplier:  Seracare Life Sciences Inc 53100324
Encompass_Preferred
Affinity purified polyclonal antibody to the Campylobacter species made in Goat. Isolated from a serum pool of goats immunized with different strains of heat-killed whole cells of Camplyobacter. This antibody is highly specific for Camplyobacter.
Catalog No. 50-668-61