Learn More
Immunology Consultants anti-Canine Heartworm Antibody (Chicken) - HRP Conjugated
SDP
Catalog No. 5000435305
Encompass_Preferred
This HRP‑conjugated anti‑Canine Heartworm antibody provides a powerful enzyme‑based detection reagent for heartworm antigens in ELISA, Western blotting, and chemiluminescent assays. Produced in chicken and supplied at 1 mg, this conjugate enables direct HRP detection without secondary antibodies, improving workflow efficiency and reducing background. Its strong specificity for Dirofilaria immitis antigens makes it ideal for diagnostic assay development, antigen screening, and veterinary infectious disease research. A dependable, high‑performance tool for sensitive heartworm detection.
Catalog No. 50-004-35305 Supplier Immunology Consultants Supplier No. CHRT65P
This HRP‑conjugated anti‑Canine Heartworm antibody provides a powerful enzyme‑based detection reagent for heartworm antigens in ELISA, Western blotting, and chemiluminescent assays. Produced in chicken and supplied at 1 mg, this conjugate enables direct HRP detection without secondary antibodies, improving workflow efficiency and reducing background. Its strong specificity for Dirofilaria immitis antigens makes it ideal for diagnostic assay development, antigen screening, and veterinary infectious disease research. A dependable, high‑performance tool for sensitive heartworm detection.