Learn More
Immunology Consultants Affinity Purified Chicken anti-Canine Leptospira Lig A/B Antibody, 1.0 mg
SDP
Catalog No. NC2734277
Encompass_Preferred
Chickens were immunized with a highly purified protein corresponding to a conserved sequence of the canine Leptospira immunoglobulin (Ig)-like protein (Lig), and the eggs were collected and delipidated. Antibody was immunoaffinity purified off a canine Leptospira Lig protein containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. This antibody reacts with canine Leptospira Lig protein and is suitable for blotting, ELISA and IHC applications. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. NC2734277 Supplier Immunology Consultants Supplier No. CLAB40A
Chickens were immunized with a highly purified protein corresponding to a conserved sequence of the canine Leptospira immunoglobulin (Ig)-like protein (Lig), and the eggs were collected and delipidated. Antibody was immunoaffinity purified off a canine Leptospira Lig protein containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. This antibody reacts with canine Leptospira Lig protein and is suitable for blotting, ELISA and IHC applications. Optimal working dilutions should be determined experimentally by the investigator.