Learn More
Immunology Consultants Affinity Purified Chicken anti-Canine Leptospira LipL32 Antibody, 1.0 mg
SDP
Catalog No. NC2734276
Encompass_Preferred
Chickens were immunized with highly purified canine Leptospira LipL32 protein and the eggs were collected and delipidated. Antibody was immunoaffinity purified off a canine Leptospira LipL32 protein containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. This antibody reacts with canine Leptospira LipL32 protein and is suitable for blotting, ELISA and IHC applications. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. NC2734276 Supplier Immunology Consultants Supplier No. CL3240A
Chickens were immunized with highly purified canine Leptospira LipL32 protein and the eggs were collected and delipidated. Antibody was immunoaffinity purified off a canine Leptospira LipL32 protein containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.32 equals 1.0 mg of IgY. This antibody reacts with canine Leptospira LipL32 protein and is suitable for blotting, ELISA and IHC applications. Optimal working dilutions should be determined experimentally by the investigator.