Learn More
Ted Pella Inc DUMONT Biology Anti-Capillary Reverse (self-closing) Tweezers Biology Grade 5ACN 108mm NM-SS
SDP
Catalog No. NC0088618
Encompass
These anti-capillary, reverse (self closing) and regular tweezers are great for handling transmission electron microscopy grids. The picture above shows the anti-capillary tweezer holding an electron microscopy grid. The tweezers tips are shaped and aligned to pick up less moisture when removing grids from a liquid. The top (bent) tip meets the bottom(flat) tip at a slight distance behind the point, thus forming a small lip which supports the grid and allows it to be picked up without twisting.
Catalog No. NC0088618 Supplier Ted Pella Inc Supplier No. 5105NM
These anti-capillary, reverse (self closing) and regular tweezers are great for handling transmission electron microscopy grids. The picture above shows the anti-capillary tweezer holding an electron microscopy grid. The tweezers tips are shaped and aligned to pick up less moisture when removing grids from a liquid. The top (bent) tip meets the bottom(flat) tip at a slight distance behind the point, thus forming a small lip which supports the grid and allows it to be picked up without twisting.