Learn More
Immunology Consultants anti-Carcinoembryonic Antigen (CEA) Antibody (Clone #104)
SDP
Catalog No. 5000435538
Encompass_Preferred
Clone 104 targets a distinct epitope on CEA, enabling precise, high‑specificity detection in immunoassays. Its strong affinity supports ELISA, Western blotting, and tissue‑based applications, making it valuable for oncology research, tumor profiling, and diagnostic assay development. MCEA‑80A‑104 provides clean, reproducible signal across serum, cell lysates, and purified antigen preparations, ensuring dependable performance in biomarker studies and assay validation workflows
Catalog No. 50-004-35538 Supplier Immunology Consultants Supplier No. MCEA80A104
Clone 104 targets a distinct epitope on CEA, enabling precise, high‑specificity detection in immunoassays. Its strong affinity supports ELISA, Western blotting, and tissue‑based applications, making it valuable for oncology research, tumor profiling, and diagnostic assay development. MCEA‑80A‑104 provides clean, reproducible signal across serum, cell lysates, and purified antigen preparations, ensuring dependable performance in biomarker studies and assay validation workflows