Learn More
Bioworld Anti-Caspase 12 Antibody (Rabbit Polyclonal IgG), 200 μg
Supplier:  Bioworld 203121171
Encompass_Preferred
Anti-Caspase 12 Antibody (Rabbit Polyclonal IgG)Cat#20312117DetailsHost:RabbitIsotype:IgGClass:PolyclonalImmunogen:Synthetic peptide corresponding to Human, Mouse, and Rat Caspase 12 (Propeptide and cleaved forms)Conjugate:UnconjugatedForm InformationForm:LiquidConcentration:2 mg/mLPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:Store at +4°C short term (1-2 weeks). Aliquot and store at -20°C or -80°C. Avoid repeated freeze / thaw cycles.Usage RecommendationsWestern Blot (WB):1:3000-1:5000ELISA:1:150-1:400Immunoprecipitation (IP):1:350 - 1:500
Catalog No. 50-198-043