Learn More
ECM Bioscience Caspase-8 (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CM3821
Encompass_Preferred
Clone (M382) was generated from a recombinant protein containing amino acid residues in the C-terminal region of human Caspase-8. Accession #Q14790. The antibody detects a 55 kDa* protein corresponding to the molecular weight of Caspase-8 on SDS-PAGE immunoblots of human Jurkat, K562, and A431 cells. Applications: WB, E; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Caspase-8, ced3, NEDD2-like protein,p15, p10
Catalog No. 50-206-8669