Learn More
Seracare Life Sciences Inc ANTI-CAT IGG (GAMMA) AB FITC
Supplier:  Seracare Life Sciences Inc 52300315
Encompass_Preferred
Affinity purified polyclonal antibody to Cat IgG, heavy chains (gamma) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-669-01