Learn More
Seracare Life Sciences Inc ANTI-CAT IGG (H+L) AB AP
Supplier:  Seracare Life Sciences Inc 52200379
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Cat IgG, heavy and light chains (whole IgG) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-49