Learn More
Seracare Life Sciences Inc ANTI-CAT IGM (MU) AB FITC
Supplier:  Seracare Life Sciences Inc 52300316
Encompass_Preferred
Affinity purified polyclonal antibody to Cat IgM, heavy chains (mu) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-669-02