Learn More
Seracare Life Sciences Inc ANTI-CAT IGM (MU) AB HRP
Catalog No. 5066947
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Cat IgM, heavy chains (mu) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-669-47 Supplier Seracare Life Sciences Inc Supplier No. 52200292
Affinity purified polyclonal secondary antibody against Cat IgM, heavy chains (mu) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.