Learn More
Immunology Consultants anti-Cat SAA Antibody (Mouse - Clone #3A5) - Monoclonal
SDP
Catalog No. 5000435408
Encompass_Preferred
Clone 3A5 targets a distinct feline SAA epitope, providing complementary detection to Clone 1C12. This monoclonal antibody supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MSAA‑20A‑3A5 is well suited for diagnostic assay development, inflammation research, and monitoring acute‑phase responses in cats. Its reproducible performance ensures dependable detection across diverse sample matrices.
Catalog No. 50-004-35408 Supplier Immunology Consultants Supplier No. MSAA20A3A5
Clone 3A5 targets a distinct feline SAA epitope, providing complementary detection to Clone 1C12. This monoclonal antibody supports sensitive measurement in ELISA, Western blotting, and antigen‑capture workflows, offering strong affinity and minimal nonspecific binding. MSAA‑20A‑3A5 is well suited for diagnostic assay development, inflammation research, and monitoring acute‑phase responses in cats. Its reproducible performance ensures dependable detection across diverse sample matrices.