Learn More
Immunology Consultants anti-Cat SAA Antibody (Mouse - Clone #4B2) - Monoclonal
SDP
Catalog No. 5000435457
Encompass_Preferred
This monoclonal antibody recognizes a unique epitope on feline SAA, enabling precise detection in immunoassays. Clone 4B2 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture formats. MSAA‑20A‑4B2 is ideal for veterinary diagnostics, biomarker development, and studies examining feline inflammatory responses. Its consistent lot‑to‑lot performance ensures reliable detection across repeated assays.
Catalog No. 50-004-35457 Supplier Immunology Consultants Supplier No. MSAA20A4B2
This monoclonal antibody recognizes a unique epitope on feline SAA, enabling precise detection in immunoassays. Clone 4B2 delivers strong affinity and low background in ELISA, Western blotting, and antigen‑capture formats. MSAA‑20A‑4B2 is ideal for veterinary diagnostics, biomarker development, and studies examining feline inflammatory responses. Its consistent lot‑to‑lot performance ensures reliable detection across repeated assays.