Learn More
Sino Biological Anti-Cathepsin B Antibody, Rabbit Polyclonal 100 µL
Catalog No. 5000750637
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Cathepsin B (rh Cathepsin B; Catalog#10483-H08H; NP_001899.1; Arg18-Ile339). Cathepsin B specific IgG was purified by Human Cathepsin B affinity chromatography.
Catalog No. 50-007-50637 Supplier Sino Biological Supplier No. 10483T08100
Produced in rabbits immunized with purified, recombinant Human Cathepsin B (rh Cathepsin B; Catalog#10483-H08H; NP_001899.1; Arg18-Ile339). Cathepsin B specific IgG was purified by Human Cathepsin B affinity chromatography.