Learn More
Selleck Chemical LLC Anti-CCL2-InVivo-A2132-1MG
SDP
Catalog No. 503135913
Encompass_Preferred
Anti-mouse/human/rat CCL2-InVivo (Clone 2H5) antibody reacts with mouse human and rat CCL2 also known as monocyte chemoattractant protein 1 MCP-1 is a 13 kDa chemokine produced mainly by monocytes/macrophages and is a potent monocyte-attracting chemokine The 2H5 antibody has been shown to neutralize the biological effects of CCL2 in vivo
Catalog No. 50-313-5913 Supplier Selleck Chemical LLC Supplier No. A21321MG
Anti-mouse/human/rat CCL2-InVivo (Clone 2H5) antibody reacts with mouse human and rat CCL2 also known as monocyte chemoattractant protein 1 MCP-1 is a 13 kDa chemokine produced mainly by monocytes/macrophages and is a potent monocyte-attracting chemokine The 2H5 antibody has been shown to neutralize the biological effects of CCL2 in vivo