Learn More
ABclonal Technology CCNG1 Polyclonal Antibody, 100UL, Validated in:WB, IHC, IF
SDP
Supplier:  ABclonal Technology A5292
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human Cyclin G1 (NP_004051.1).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC•Swiss Prot: P51959•Gene ID:900•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-7743