Learn More
American Research Products Inc Anti-CD233/(Band 3) extracellular domain Monoclonal Antibody, FITC (50 Tests)
Supplier:  American Research Products Inc 0894393
Encompass
FITC-conjugate, in PBS buffer, pH 7.2 containing 0.1% sodium azide, 1% BSA. Storage: 2-8C, do not freeze
Catalog No. NC1433740