Learn More
Beckman Coulter CD127-PE, R34.34, 2 mL, ASR
Supplier:  Beckman Coulter IM1980U
Encompass
The CD127 antigen is the human IL-7 receptor alpha chain. It is a 75-80 kDa molecule. CD127 is found on normal B cell precursors but not on mature B lymphocytes. It is also expressed by the majority of T lymphocytes, a subset of monocytes and a subset of CD34+ cells. CD127 associates with CD132 (IL-2Rgamma) to form the high affinity IL-7 receptor. Receptor expression is down-regulated following T cell activation.
Catalog No. NC1696662