Learn More
Biolegend APC/Fire™ 750 anti-human CD138 (Syndecan-1)
Supplier:  Biolegend 352316
Encompass_Preferred
APC/Fire™ 750 anti-human CD138 (Syndecan-1) [DL-101]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. NC1676117