Learn More
ECM Bioscience CD147/Emmprin/Basigin (Extracellular region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CM0521
Encompass_Preferred
Clone (M052) was generated from a proprietary antigen related to the native human CD147 expressed in NCI-H1915 lung cancer cell line. Accession #P35613. Clone M052 mouse monoclonal antibody detects a 38-58 kDa* protein on SDS-PAGE immunoblots of human SK-MES1, MDA-MB-231, MeWo, A431, LNCaP, and NCI-H1915 cells, as well as human brain tissues. The antibody works for western blot and ELISA detection of CD147. Applications: WB, E, ; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Basigin, 5F7, Collagenase stimulatory factor, Extracellular matrix metalloproteinase,EMMPRIN, Leukocyte activation antigen M6, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor, TCSF, CD147, BSG
Catalog No. 50-206-8646