Learn More
BPS Bioscience Inc Anti-CD19 CAR-T Cells, 5PK, 2 x 10^6 cells per vial, 1 ml per vial

Supplier: BPS Bioscience Inc 781712
The BPS Bioscience Anti-CD19 CAR-T Cells (5PK) are engineered chimeric antigen receptor (CAR) T cells designed for research applications involving CD19-targeted therapies. These cells are created using high-titer lentiviral transduction of human CD4+CD8+ T cells and validated for CAR expression, providing a reliable tool for cytotoxicity assays and modulator screening.
- Target antigen: CD19
- Cryopreservation medium: CryoStor CS10
- Quantity: 5 vials
- Cells per vial: 2 x 10^6 cells
- Volume per vial: 1 ml
- Application: Positive control, modulator screening, cytotoxicity assay optimization
- Shipping condition: Dry ice
- Long-term storage: Liquid nitrogen
- Research use only: Yes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.