Learn More
BPS Bioscience Inc Anti-CD19 CAR / NFAT (Luciferase) Reporter Jurkat Cell Line, 2 vials
SDP
Supplier:  BPS Bioscience Inc 79853
Encompass
The Anti-CD19 CAR / NFAT (Luciferase) Reporter Jurkat Cell Line is a specialized research tool designed for evaluating anti-CD19 CAR constructs and lentiviruses. This double stable Jurkat cell line integrates a robust CAR design with an NFAT-dependent luciferase reporter, enabling efficient measurement of CAR activation in co-culture with CD19-expressing target cells. It streamlines the validation of CAR T-cell development workflows through its comprehensive design and validated performance.
  • Comprehensive CAR design with anti-CD19 scFv linked to a 3rd generation CAR
  • NFAT-luciferase reporter system for quantitative CAR activation assessment
  • Validated for CAR expression using FACS and luciferase activation
  • Ideal for primary screening and functional validation of CAR constructs
  • Screened for mycoplasma absence
  • Stored at -80°C
  • BSL-2 biosafety level
  • Research use only
  • Supplied in 2 vials
Catalog No. NC3450048
Add to Cart
Request a Quote