Learn More
ECM Bioscience CD244/2B4/SLAMF4 (Extracellular region) M053 Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CM0531
Encompass_Preferred
Clone (M053) was generated from a recombinant protein that included the extracellular region of human CD244 protein. Accession #Q9BZW8. Clone M053 mouse monoclonal antibody detects a 110 kDa* protein on SDS-PAGE "Native" or denatured immunoblots of human THP1 monocytes. The antibody prefers the native CD244 protein, but also detects denatured CD244. The antibody works for western blot, immunoprecipitation, immunocytochemistry, and ELISA capture. Applications: WB, EC, IP, ICC; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = Natural killer cell receptor 2B4, 2B4, NK cell activation-inducing ligand, NAIL, NK cell type I receptor protein 2B4, NKR2B4, h2B4, SLAM family member 4, SLAMF4, Signaling lymphocytic activation molecule 4, CD244
Catalog No. 50-206-8647