Learn More
American Research Products Inc Anti-CD31 Monoclonal Antibody, Specific For Mouse CD31, Validated By Over 185 Citations, 500 µl
Supplier:  American Research Products Inc DIA310
Encompass
The Anti-CD31 Monoclonal Antibody, clone SZ31, enables specific detection of murine CD31 in FFPE tissue sections. Identified for high-quality immunohistochemical analysis, this antibody addresses common limitations in standard formalin-fixed samples. Its unique focus on murine models enhances research in cell adhesion and angiogenesis without cross-reacting with human CD31, backed by over 185 validations.
Catalog No. NC2028563